Merck
DWK217313602
DURAN® wide mouth reaction flask with beaded rim
round bottom glass flask, flask capacity (250 mL), single-neck
Synonym(e):
217313602
Größe auswählen
10 EACH
105,00 €
105,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EACH
105,00 €
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.17
105,00 €
Check Cart for Availability
Materialien
round bottom glass flask
Verpackung
case of 10 ea
Hersteller/Markenname
DURAN® 217313602
Kolbenkapazität
(250 mL)
Allgemeine Beschreibung
Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)