Merck
CLS36060600F
Pyrex® Buchner funnel with fritted disc
capacity 600 mL, porosity 4-5.5 μm
Synonym(e):
Corning® 36060
About This Item
UNSPSC-Code:
41121809
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Hersteller/Markenname
Corning 36060-600F
Kapazität
600 mL
Scheibendurchm.
90 mm
Porengröße
4-5.5 μm porosity
Allgemeine Beschreibung
Especially useful where filter paper would be attacked by filtered material. Disc should not be subjected to pressure differential in excess of 1 atm. Top edge is beaded. Granular frit.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)