Merck
BR782880
BRAND® BRANDplates® for inserts
24-well with lid cellGrade S
Synonym(e):
BRANDplates®, Cell culture, Insert System, Inserts, Plates, Tissue culture
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
polystyrene
transparent wells
Sterilität
sterile
Leistungsmerkmale
lid
single packed
Verpackung
pack of 10 ea
pkg of Individually wrapped, with lid
Hersteller/Markenname
BRAND 782880
Größe
24 well
Verwandte Kategorien
Allgemeine Beschreibung
PS. pureGrade S surface, untreated, sterile, cover with condensation rings.Manufactured according to the requirements in ANSI/SLAS Standards 1 and 4.BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Leistungsmerkmale und Vorteile
• Non-treated surface• Side well access for easier pipetting and removing cover slips• Perfect positioning of the BRANDplates® insert
Rechtliche Hinweise
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG