Merck
BR703980
BRAND® Software for PLT unit
version 2.05.002
Größe auswählen
1 EA
140,00 €
140,00 €
Versandbereit am22. April 2025Details
Größe auswählen
Ansicht ändern
1 EA
140,00 €
About This Item
UNSPSC-Code:
41122804
NACRES:
NB.43
140,00 €
Versandbereit am22. April 2025Details
Leistungsmerkmale
version 2.05.002
Hersteller/Markenname
BRAND 703980
Verwandte Kategorien
Allgemeine Beschreibung
The PLTconnect software gives you the additional security of knowing all your test results are documented at all times. PLTconnect transmits test results for each pipette automatically and securely through the USB port on the PC, storing them there in a database or as a test certificate. This means you always have access to test results for your pipettes, and can print out certificates as needed
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)