Merck
BR701460
BRAND® Reservoir with cover
polypropylene, capacity 40 mL
Größe auswählen
1 EACH
91,80 €
91,80 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EACH
91,80 €
About This Item
UNSPSC-Code:
41121517
NACRES:
NB.01
91,80 €
Check Cart for Availability
Materialien
polypropylene
Hersteller/Markenname
BRAND 701460
Kapazität
40 mL
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Reservoir with Cover is designed for both manual pipetting and automation, providing a reliable solution for liquid handling in laboratory settings. Made from durable polypropylene (PP), this reservoir has a working volume of 40 ml and features a V-bottom design that minimizes dead volume, ensuring efficient liquid use. The graduated markings allow for precise measurement, enhancing accuracy in experiments.CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)