Merck
BR4630141
BRAND® Dispensette® S Organic, Analog-adjustable bottle-top dispenser
volume range 1-10 mL, with recirculation valve
Synonym(e):
bottle top dispensers
Größe auswählen
1 EA
404,00 €
404,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
404,00 €
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
404,00 €
Check Cart for Availability
Leistungsmerkmale
incremental 0.2 mL
DE-M marking
with recirculation valve
Hersteller/Markenname
BRAND 4630141
Volumenbereich
1-10 mL
Fitting
(GL 28/S28)
GL 32
GL 38
S 40
Allgemeine Beschreibung
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Leistungsmerkmale und Vorteile
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Sonstige Hinweise
Analog-adjustable - Rapid and safe volume setting with new internal scalloped track. Simple calibration adjustment with a supplied tool.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Protokolle
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Verwandter Inhalt
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.