Merck
BR136110
BRAND® stirring spatula
polypropylene
Synonym(e):
Stirring rods
Größe auswählen
10 EA
19,00 €
19,00 €
Versandbereit am14. April 2025Details
Größe auswählen
Ansicht ändern
10 EA
19,00 €
About This Item
UNSPSC-Code:
41122403
NACRES:
NB.43
19,00 €
Versandbereit am14. April 2025Details
Materialien
polypropylene
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 136110
L × T
245 mm × 6 mm
Allgemeine Beschreibung
The BRAND® stirring spatula is made from durable polypropylene (PP) and features two differently shaped ends, each approximately 15 mm wide. This versatile design allows for effective mixing, stirring, and scraping of various substances in laboratory applications. The lightweight construction makes it easy to handle, while the chemical resistance of PP ensures durability against a range of reagents.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)