Merck
BR135820
BRAND® stirring rod
AR-Glass, L × D 250 mm × 8 mm
Synonym(e):
Stirring rods
Größe auswählen
50 EA
56,00 €
56,00 €
Versandbereit am28. April 2025Details
Größe auswählen
Ansicht ändern
50 EA
56,00 €
About This Item
UNSPSC-Code:
41121802
NACRES:
NB.72
56,00 €
Versandbereit am28. April 2025Details
Materialien
AR-Glass
Beschreibung
fused closely at both ends
Verpackung
pack of 50 ea
Hersteller/Markenname
BRAND 135820
L × T
250 mm × 8 mm
Allgemeine Beschreibung
The BRAND® stirring rod is a glass tube that is closed on both ends, providing a reliable and durable tool for mixing and stirring various solutions in laboratory settings. Its smooth surface ensures easy cleaning and maintenance, while the closed design prevents contamination.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)