Merck
BMSW1100200
Sigma-Aldrich
Accuris F1 Grade Calibration Weight
weight set
Synonym(e):
milligram calibration weight, top pan balance weight, top pan calibration
Größe auswählen
1 EA
584,00 €
584,00 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
584,00 €
About This Item
UNSPSC-Code:
41111502
NACRES:
NB.71
584,00 €
Check Cart for Availability
Hersteller/Markenname
Benchmark Scientific W1100-200
Allgemeine Beschreibung
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Precision, compact, and portable balances (readability 0.1 to 0.001g) can be calibrated with our Basic Class F1 Calibration Set. Included in the set are 23 stainless steel cylindrical and leaf weights: 2 x 200g, 1 x 100g, 1 x 50g, 2 x 20g, 1 x 10g, 1 x 5g, 2 x 2g, 1 x 1g, 1 x 500mg, 2 x 200mg, 1 x 100mg, 1 x 50mg, 2 x 20mg, 1 x 10mg, 1 x 5mg, 2 x 2mg, 1 x 1mg. Cylindrical weights are marked with their nominal value. All are supplied in a protective case along with tweezers, a cleaning cloth and dust brush.
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Fragen
  1. Does the calibration set come with a certificate of analysis (for what the actual weight is of each weight)?
    1 Antwort
    1. Yes, each individual weight and weight set from Accuris is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
      Hilfreich?