BMSBR5000EUK
Everlast™ rocker with flat mat
AC/DC input 230 V AC, UK plug
Größe auswählen
1.380,00 €
Größe auswählen
About This Item
1.380,00 €
Empfohlene Produkte
Hersteller/Markenname
Benchmark Scientific BR5000-E-UK
B × T
16 in. × 12 in.
Verwandte Kategorien
Allgemeine Beschreibung
- Stainless-steel platform
- Dual platform accessory doubles work surface
- Cold room and incubator safe
The rocking speed and tilt angle can easily be adjusted
to vary between gentle and aggressive agitation. Both
the standard platform and the (optional) stacking
platform are constructed of stainless steel, preventing
against any rusting or corrosion. The Everlast 247 is
designed to provide years of uninterrupted service.
Rechtliche Hinweise
Ähnliches Produkt
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)