Merck
BAH195610000
SP Bel-Art® Straight Tubing Connectors
tubing I.D. 0.375-0.375 in. (6-9.5 mm)
Synonym(e):
connectors, tubing, tubing coupler
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Verpackung
pack of 12 ea
Hersteller/Markenname
Bel-Art H19561-0000
L × B
1.625 in. (41.3 mm) × 0.4375 in. (11.1 mm)
Schlauch ID
0.375-0.375 in. (6-9.5 mm)
Allgemeine Beschreibung
These straight fittings ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.).
Leistungsmerkmale und Vorteile
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases.
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)