Merck
BAF187990000
Scienceware® test tube capper
Polypropylene with vinyl covered handhold
Größe auswählen
1 EA
65,60 €
65,60 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
65,60 €
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
65,60 €
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F18799-0000
Durchm. × H
4.4 cm × 8.9 cm
Allgemeine Beschreibung
Eliminates fatigue and soreness of hands and fingers from the repetitive task of applying test tube caps. Test tube capper fits comfortably in the palm of your hand. Use tip to apply pressure and securely cap tubes.
  • Capping can be performed with test tubes in a rack or while being held in the hand
  • Designed for use with recessed center test tube caps
  • Polypropylene with vinyl covered handhold
Rechtliche Hinweise
Scienceware is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)