BAF131600009
Scienceware® biohazard disposal bags
width × H × thickness 300 mm (12 in.) × 24 in. (610 mm) × 1.5 mil (0.04 mm), pack of 100 ea, translucent polypropylene
Synonym(e):
Scienceware® biohazard disposal bags
About This Item
Empfohlene Produkte
Materialien
translucent polypropylene
Leistungsmerkmale
label (With Biohazard Warning Label and Sterilization Indicator Patch)
Verpackung
pack of 100 ea
Hersteller/Markenname
Bel-Art F13160-0009
Kapazität
3.8-11.4 L (1-3 gal)
width × H × thickness
300 mm (12 in.) × 24 in. (610 mm) × 1.5 mil (0.04 mm)
Allgemeine Beschreibung
Leistungsmerkmale und Vorteile
- Transparent polypropylene; 0.03mm (1.5 mil) thick
- Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
- High temperature autoclavable at a maximum of 135?C (275?F)
- Plain, non-printed bags can also be used as conventional trash can liners
- Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
- Bags with biohazard warning label contain standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
- Transparent polypropylene; 0.03mm (1.5 mil) thick
- Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
- High temperature autoclavable at a maximum of 135°C (275°F)
- Plain, non-printed bags can also be used as conventional trash can liners
- Sterilization indicator patch darkens when exposed to steam sterilization for 20 minutes
- Bags with biohazard warning label contain standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)