Merck
  • Fluorometric immunocapture assay for the specific measurement of matrix metalloproteinase-9 activity in biological samples: application to brain and plasma from rats with ischemic stroke.
Fluorometric immunocapture assay for the specific measurement of matrix metalloproteinase-9 activity in biological samples: application to brain and plasma from rats with ischemic stroke.
Molecular brain (2013-03-26)
Kimberly E Hawkins, Kelly M DeMars, Changjun Yang, Gary A Rosenberg, Eduardo Candelario-Jalil
ABSTRACT
Matrix metalloproteinases are important factors in the molecular mechanisms leading to neuronal injury in many neurological disorders. Matrix metalloproteinase (MMP)-9 is up-regulated after cerebral ischemia and neuroinflammation and is actively involved in blood-brain barrier disruption. Current methods of measuring MMP-9 activity, such as gelatin-substrate zymography, are unspecific and arduous. Here we developed an immunocapture assay with high efficiency, specificity, and sensitivity for quantifying endogenously active as well as total MMP-9 activity. A fluorescence resonance energy transfer (FRET) peptide-based immunocapture assay was developed that enables the accurate assessment of total and active forms of MMP-9 in complex biological samples. The FRET assay demonstrated correct and efficient binding of MMP-9 to a mouse monoclonal MMP-9 antibody and high specificity of the immunocapture antibody for MMP-9. Total and active levels of MMP-9 were measured in rat brain homogenates, plasma, human HT-1080 conditioned media, and RBE4 endothelial cell lysates. The FRET immunocapture assay yielded highly similar results for total MMP-9 activity when compared to gelatin-substrate zymography. We suggest that the new FRET peptide-based immunocapture assay is a viable replacement of zymography for sensitive and high throughput quantification of MMP-9 activity in biological samples.
Sigma-Aldrich
Gelatin, tested according to Ph. Eur.
Sigma-Aldrich
Gelatin from bovine skin, Type B
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
(R)-N4-Hydroxy-N1-[(S)-2-(1H-indol-3-yl)-1-methylcarbamoyl-ethyl]-2-isobutyl-succinamide, >95% (HPLC)
Sigma-Aldrich
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
Millipore
Gelatin from porcine skin, suitable for microbiology, low gel strength
Millipore
Gelatin from porcine skin, medium gel strength, suitable for microbiology
Millipore
Gelatin from porcine skin, suitable for microbiology, high gel strength
Sigma-Aldrich
4-Aminophenylmercuric acetate, ≥90% (titration)
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Gelatin from porcine skin, gel strength 300, Type A
Sigma-Aldrich
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
Sigma-Aldrich
Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
SB−3CT, ≥98% (HPLC), powder
Sigma-Aldrich
Gelatin from cold water fish skin, solid
Sigma-Aldrich
Gelatin from porcine skin, gel strength 80-120 g Bloom, Type A
Sigma-Aldrich
Prionex® Highly purified Type A, aqueous solution
Millipore
Gelatin from porcine skin, suitable for microbiology, ultrahigh gel strength