Skip to Content
Merck

Skip To

V900392

D-(+)-Glucose

Vetec, reagent grade, ≥99.5% (HPLC)

Synonym(s):

Dextrose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
UNSPSC Code:
12352201
PubChem Substance ID:
EC Number:
200-075-1
Beilstein/REAXYS Number:
1724615
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

product line

Vetec

assay

≥99.5% (HPLC)

form

powder

technique(s)

HPLC: suitable

color

white

mp

150-152 °C (lit.)

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

General description

D-(+)-Glucose, a major product of photosynthesis, is a monosaccharide, which is a primary source of energy in the form of ATP for living organisms. It is the main component of starch (as stored energy) and cellulose (constituent of cell walls) found in the plants.

Application

D-(+)-Glucose can be used:
  • In the investigation of glucose sensor performance.[1]
  • As an initiator in Butlerov reaction.
  • In the preparation of an acid catalyst, which is used in the alkylation of phenols.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under D-(+)-Glucose


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters