Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
292.24
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
200-449-4
Beilstein/REAXYS Number:
1716295
MDL number:
eCl@ss:
39030907
Pricing and availability is not currently available.
Product Name
Ethylenediaminetetraacetic acid, Vetec™, reagent grade, 98%
grade
reagent grade
product line
Vetec™
assay
98%
form
powder or crystals
mp
250 °C (dec.) (lit.)
solubility
1 M NaOH: 1 g/10 mL, clear, colorless
density
1.46 g/cm3 at 20 °C
functional group
amine, carboxylic acid
SMILES string
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
InChI
1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)
InChI key
KCXVZYZYPLLWCC-UHFFFAOYSA-N
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Ethylenediaminetetraacetic acid
signalword
Warning
hcodes
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
target_organs
Respiratory Tract
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Irrit. 2 - STOT RE 2 Inhalation
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lucienne Tritten et al.
PLoS neglected tropical diseases, 8(7), e2971-e2971 (2014-07-18)
Filarial nematodes cause chronic and profoundly debilitating diseases in both humans and animals. Applications of novel technology are providing unprecedented opportunities to improve diagnosis and our understanding of the molecular basis for host-parasite interactions. As a first step, we investigated
Khosrow Adeli et al.
Clinical chemistry, 61(8), 1063-1074 (2015-06-06)
Defining laboratory biomarker reference values in a healthy population and understanding the fluctuations in biomarker concentrations throughout life and between sexes are critical to clinical interpretation of laboratory test results in different disease states. The Canadian Health Measures Survey (CHMS)
Cecilia Lezcano et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 27(9), 1193-1202 (2014-01-18)
Of more than 150 000 published studies evaluating new biomarkers, fewer than 100 biomarkers have been implemented for patient care. One reason for this is lack of rigorous testing by the medical community to validate claims for biomarker clinical relevance, and

