Skip to Content
Merck

Skip To

1606026

USP

Rosuvastatin Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(Calcium (3R,5S,E)-7-{4-(4-fluorophenyl)-2-[(2-hydroxy-N,2-dimethylpropyl)sulfonamide]-6-isopropylpyrimidin-5-yl}-3,5-dihydroxyhept-6-enoate salt (1:2))

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
Ca(C25H33FN3O7S)2
CAS Number:
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

rosuvastatin

manufacturer/tradename

USP

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

USP issued SDS can be found here.

Application

Rosuvastatin Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
160603711632641604552
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

-

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

agency

USP

agency

USP

agency

-

agency

-

API family

rosuvastatin

API family

rosuvastatin

API family

dapagliflozin

API family

rivaroxaban


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT RE 2 Oral

target_organs

Liver,Kidney

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1606026-25MG04065268921852

Questions

Reviews

No rating value

Active Filters