Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 mg | Please contact Customer Service for Availability | CZK 28,800.00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
ramipril
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
N21[C@@H]3[C@H](C[C@H]2C(=O)N([C@H](C1=O)C)[C@@H](CCc4ccccc4)C(=O)OCC)CCC3
InChI
1S/C23H30N2O4/c1-3-29-23(28)19(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-18-11-7-10-17(18)14-20(25)22(24)27/h4-6,8-9,15,17-20H,3,7,10-14H2,1-2H3/t15-,17-,18-,19-,20-/m0/s1
InChI key
KOVMAAYRBJCASY-JBDAPHQKSA-N
General description
Application
- Ramipril
- Ramipril Tablets
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format neat | format neat |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. - |
| API family ramipril | API family ramipril | API family ramipril | API family trandolapril |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
