Skip to Content
Merck

Skip To

1466641

USP

N-Nitrosodibutylamine (NDBA)

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-Butyl-N-nitroso-1-butanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
CZK 15,500.00

About This Item

Empirical Formula (Hill Notation):
C8H18N2O
CAS Number:
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 15,500.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

packaging

pkg of 1 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecules)

format

neat

storage temp.

−20°C

SMILES string

N(N=O)(CCCC)CCCC

InChI

1S/C8H18N2O/c1-3-5-7-10(9-11)8-6-4-2/h3-8H2,1-2H3

InChI key

YGJHZCLPZAZIHH-UHFFFAOYSA-N

General description

Nitrosamines are chemical compounds that can form during drug manufacturing and may pose health risks, including carcinogenicity.

This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including certificate/ product information sheets, please go to the website of the issuing Pharmacopoeia.

Application

N-Nitrosodibutylamine (NDBA) USP reference standard is intended for use only as specifically prescribed in the United States Pharmacopoeia.

It is also used to prepare standard, nitrosamine standards stock solution mixture, and sensitivity solutions to determine NDBA impurity in drug substances and drug products (valsartan, irbesartan, and losartan potassium etc.) by chromatography according to general chapter <1469> of United States Pharmacopeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR360814666741466652
application(s)

pharmaceutical (small molecules)

application(s)

pharmaceutical

application(s)

USP Biologics
pharmaceutical (small molecules)

application(s)

USP Biologics
pharmaceutical (small molecules)

format

neat

format

-

format

single component solution (solution in methanol)

format

neat

grade

pharmaceutical primary standard (

)

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard (

)

grade

pharmaceutical primary standard (

)

storage temp.

−20°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C

packaging

pkg of 1 mg

packaging

pkg of 1 mL

packaging

pkg of 1 mg

packaging

pkg of 1 mg

manufacturer/tradename

USP

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

manufacturer/tradename

USP

manufacturer/tradename

USP


Still not finding the right product?

Explore all of our products under N-Nitrosodibutylamine (NDBA)


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F

flash_point_c

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters