Skip to Content
Merck

Skip To

1463927

USP

Nisoldipine

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
250 mg
Please contact Customer Service for Availability
CZK 51,300.00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

CZK 51,300.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

nisoldipine

form

solid

manufacturer/tradename

USP

application(s)

pharmaceutical

storage temp.

−20°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nisoldipine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
146415814641831464169
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical

form

solid

form

solid

form

solid

form

solid

API family

nisoldipine

API family

nisoldipine

API family

nisoldipine

API family

nisoldipine

storage temp.

−20°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1463927-250MG04061841963714

Questions

Reviews

No rating value

Active Filters