Skip to Content
Merck

Skip To

1445392

USP

Modafinil sulfone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-[(Diphenylmethyl)sulfonyl]acetamide, 2-(Benzhydrylsulfonyl)acetamide, Modafinil sulfone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
30 mg
Please contact Customer Service for Availability
CZK 29,100.00

About This Item

Empirical Formula (Hill Notation):
C15H15NO3S
CAS Number:
Molecular Weight:
289.35
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
9199801
MDL number:

CZK 29,100.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

modafinil

packaging

pkg of 30 mg

manufacturer/tradename

USP

drug control

Új pszichoaktív anyag / New psychoactive substance (Hungary), 78/2022. (XII. 28.) BM rendelet

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)CS(=O)(=O)C(c1ccccc1)c2ccccc2

InChI

1S/C15H15NO3S/c16-14(17)11-20(18,19)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H2,16,17)

InChI key

ZESNOWZYHYRSRY-UHFFFAOYSA-N

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Modafinil sulfone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
104275817110641445368
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

packaging

pkg of 30 mg

packaging

pkg of 500 mg

packaging

pkg of 25 mg

packaging

-

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

API family

modafinil

API family

modafinil

API family

vardenafil

API family

modafinil


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters