Skip to Content
Merck

Skip To

1443759

USP

Mifepristone

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

11β-(4-Dimethyl­amino)­phenyl-17β-hydroxy-17-(1-propynyl)­estra-4,9-dien-3-one, RU-38486, RU-486

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
CZK 14,000.00

About This Item

Empirical Formula (Hill Notation):
C29H35NO2
CAS Number:
Molecular Weight:
429.59
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CZK 14,000.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

mifepristone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CCC3=C1[C@H](C[C@@]4(C)[C@@]2([H])CC[C@@]4(O)C#CC)c5ccc(cc5)N(C)C

InChI

1S/C29H35NO2/c1-5-15-29(32)16-14-26-24-12-8-20-17-22(31)11-13-23(20)27(24)25(18-28(26,29)2)19-6-9-21(10-7-19)30(3)4/h6-7,9-10,17,24-26,32H,8,11-14,16,18H2,1-4H3/t24-,25+,26-,28-,29-/m0/s1

InChI key

VKHAHZOOUSRJNA-GCNJZUOMSA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mifepristone USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Mifepristone (RU-486) has activity as both a progesterone receptor antagonist and a glucocorticoid receptor antagonist.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M80464758381604600
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

Calbiochem®

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

-

grade

-

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

-

format

neat

API family

mifepristone

API family

-

API family

-

API family

rimexolone

Gene Information

human ... NR3C1(2908), PGR(5241)

Gene Information

human ... AR(367), ESR1(2099), ESR2(2100), HTR1A(3350), HTR1B(3351), HTR1D(3352), HTR1E(3354), HTR1F(3355), HTR2A(3356), HTR2B(3357), HTR2C(3358), HTR3A(3359), HTR3B(9177), HTR3C(170572), HTR3D(200909), HTR3E(285242), HTR4(3360), HTR5A(3361), HTR5B(645694), HTR6(3362), HTR7(3363), NR3C1(2908), NR3C2(4306), PGR(5241)
mouse ... Nr3c1(14815)
rat ... Nr3c1(24413), Tat(24813)

Gene Information

-

Gene Information

-


Still not finding the right product?

Explore all of our products under Mifepristone


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters