Mifepristone
Synonym(s):
11β-(4-Dimethylamino)phenyl-17β-hydroxy-17-(1-propynyl)estra-4,9-dien-3-one, RU-38486, RU-486, Mifepristone
Empirical Formula (Hill Notation):
C29H35NO2
CAS Number:
Molecular Weight:
429.59
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Technique
Showing 1-3 of 3