Skip to Content
Merck

Skip To

1424244

USP

Methyl caproate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl hexanoate, Caproic acid methyl ester, Hexanoic acid methyl ester, Methyl caproate

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CH3(CH2)4COOCH3
CAS Number:
Molecular Weight:
130.18
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1744683


grade

pharmaceutical primary standard

API family

methyl caproate

manufacturer/tradename

USP

density

0.884 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCC(=O)OC

InChI

1S/C7H14O2/c1-3-4-5-6-7(8)9-2/h3-6H2,1-2H3

InChI key

NUKZAGXMHTUAFE-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methyl caproate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Glyceryl Tricaprylate
  • Glyceryl Monocaprylate
  • Glyceryl Mono and Dicaprylate
  • Glyceryl Mono and Dicaprylocaprate
  • Glyceryl Monocaprylocaprate
  • Saw Palmetto Capsules
  • Powdered Saw Palmetto
  • Saw Palmetto

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
W270814215998.20638
grade

pharmaceutical primary standard

grade

FG, Halal, Kosher, natural

grade

analytical standard

grade

-

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

-

format

neat

format

-

application(s)

pharmaceutical (small molecule)

application(s)

flavors and fragrances

application(s)

-

application(s)

-

API family

methyl caproate

API family

-

API family

-

API family

-

density

0.884 g/mL at 20 °C (lit.)

density

0.885 g/mL at 25 °C (lit.)

density

0.884 g/mL at 20 °C (lit.)

density

0.88 g/cm3 at 20 °C


pictograms

Flame

signalword

Warning

hcodes

pcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

99.5 °F - closed cup

flash_point_c

37.5 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1424244-300MG04061838734334
1424244-3X100MG04065272601023

Questions

Reviews

No rating value

Active Filters