Skip to Content
Merck

Skip To

1349659

USP

Isometheptene mucate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Galactaric acid, compound with N,1,5-trimethyl-4-hexenylamine (1:2)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
CZK 12,200.00

About This Item

Empirical Formula (Hill Notation):
(C9H19N)2 · C6H10O8
CAS Number:
Molecular Weight:
492.65
MDL number:
UNSPSC Code:
41116107
EC Number:
231-315-3
NACRES:
NA.24

CZK 12,200.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

isometheptene

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N(C(CCC=C(C)C)C)C.OC(C(O)C(O)C(=O)O)C(O)C(=O)O

InChI

1S/C9H19N.C6H10O8/c1-8(2)6-5-7-9(3)10-4;7-1(3(9)5(11)12)2(8)4(10)6(13)14/h6,9-10H,5,7H2,1-4H3;1-4,7-10H,(H,11,12)(H,13,14)

InChI key

HZEDCQLNSYIOTE-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isometheptene mucate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
172702017270641013512
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

isometheptene

API family

zolmitriptan

API family

zolmitriptan

API family

almotriptan


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1349659-200MG04061838730398

Questions

Reviews

No rating value

Active Filters