Skip to Content
Merck

Skip To

1329006

USP

Hydroxyprogesterone caproate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

17α-Hydroxyprogesterone hexanoate, 17α-Hydroxyprogesterone caproate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
CZK 12,200.00

About This Item

Empirical Formula (Hill Notation):
C27H40O4
CAS Number:
Molecular Weight:
428.60
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CZK 12,200.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

hydroxyprogesterone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4(OC(=O)CCCCC)C(C)=O

InChI

1S/C27H40O4/c1-5-6-7-8-24(30)31-27(18(2)28)16-13-23-21-10-9-19-17-20(29)11-14-25(19,3)22(21)12-15-26(23,27)4/h17,21-23H,5-16H2,1-4H3/t21-,22+,23+,25+,26+,27+/m1/s1

InChI key

DOMWKUIIPQCAJU-LJHIYBGHSA-N

Gene Information

human ... PGR(5241)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydroxyprogesterone caproate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Hydroxyprogesterone Caproate Injection

Biochem/physiol Actions

Shows significant reduction of pre-term delivery by mothers with a history of premature delivery, with no detectable harm to the infant.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1378001BP221M0250000
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

Gene Information

human ... PGR(5241)

Gene Information

human ... PGR(5241)

Gene Information

human ... PGR(5241)

Gene Information

human ... PGR(5241)

API family

hydroxyprogesterone

API family

medroxyprogesterone

API family

medroxyprogesterone

API family

medroxyprogesterone


Still not finding the right product?

Explore all of our products under Hydroxyprogesterone caproate


signalword

Danger

pictograms

Health hazard

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters