Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mL | Please contact Customer Service for Availability | CZK 9,790.00 |
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
ethyl oleate
manufacturer/tradename
USP
refractive index
n20/D 1.451 (lit.)
bp
216-218 °C/15 mmHg
mp
−32 °C (lit.)
density
0.87 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
−20°C
SMILES string
CCCCCCCC\C=C/CCCCCCCC(=O)OCC
InChI
1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11-
InChI key
LVGKNOAMLMIIKO-QXMHVHEDSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| format neat | format neat | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| mp −32 °C (lit.) | mp - | mp 34-38 °C (lit.) | mp 24-26 °C (lit.) |
| density 0.87 g/mL at 25 °C (lit.) | density - | density - | density 0.857 g/mL at 25 °C (lit.) |
Still not finding the right product?
Explore all of our products under Ethyl oleate
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



