Skip to Content
Merck

Skip To

1231750

USP

Echinacoside

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
15 mg
Please contact Customer Service for Availability
CZK 7,930.00

About This Item

Empirical Formula (Hill Notation):
C35H46O20
CAS Number:
Molecular Weight:
786.73
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
4778612

CZK 7,930.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

echinacoside

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCc3ccc(O)c(O)c3)O[C@H](CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H]2OC(=O)\C=C\c5ccc(O)c(O)c5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C35H46O20/c1-14-24(42)26(44)29(47)35(51-14)55-32-30(48)34(49-9-8-16-3-6-18(38)20(40)11-16)53-22(13-50-33-28(46)27(45)25(43)21(12-36)52-33)31(32)54-23(41)7-4-15-2-5-17(37)19(39)10-15/h2-7,10-11,14,21-22,24-40,42-48H,8-9,12-13H2,1H3/b7-4+/t14-,21+,22+,24-,25+,26+,27-,28+,29+,30+,31+,32+,33+,34+,35-/m0/s1

InChI key

FSBUXLDOLNLABB-ISAKITKMSA-N

General description

Echinacoside (ECH) is a typical phenylethanoid glycoside. It is known to contain multiple unstable chemical bonds and high reactive functional groups which aids to pathways involving degradation and metabolism. It is isolated from the stems of Cistanches salsa. It is a crude drug mostly used as antisenium and antifatigue agents.

Application

Echinacoside USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Echinacea angustifolia
  • Echinacea pallida
  • Echinacea purpurea Aerial Parts
  • Echinacea purpurea Root
  • Echinacea Species Dry Extract Capsules
  • Echinacea Species Dry Extract Tablets
  • Echinacea Species Powder Capsules

Biochem/physiol Actions

Caffeic acid glycoside found in herbal medicine. Potential therapeutic for Parkinson′s disease.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0753807668PHL89188
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

PhytoLab

grade

pharmaceutical primary standard

grade

analytical standard

grade

-

grade

primary reference standard

format

neat

format

neat

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

food and beverages

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

API family

echinacoside

API family

-

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Echinacoside


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters