Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 mL | Please contact Customer Service for Availability | CZK 4,830.00 |
About This Item
grade
pharmaceutical primary standard
API family
butyl valerate
manufacturer/tradename
USP
refractive index
n20/D 1.412 (lit.)
bp
186-187 °C (lit.)
density
0.868 g/mL at 25 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
CCCCOC(=O)CCCC
InChI
1S/C9H18O2/c1-3-5-7-9(10)11-8-6-4-2/h3-8H2,1-2H3
InChI key
OKJADYKTJJGKDX-UHFFFAOYSA-N
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical |
| format neat | format neat | format neat | format neat |
| density 0.868 g/mL at 25 °C (lit.) | density 0.863 g/mL at 25 °C (lit.) | density 0.866 g/mL at 25 °C (lit.) | density 1.06 g/mL at 25 °C (lit.) |
| refractive index n20/D 1.412 (lit.) | refractive index n20/D 1.421 (lit.) | refractive index n20/D 1.416 (lit.) | refractive index n20/D 1.545 (lit.) |
Still not finding the right product?
Explore all of our products under Butyl valerate
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
152.6 °F - closed cup
flash_point_c
67 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1082672-1ML | 04061837842627 |




