Skip to Content
Merck

Skip To

1034498

USP

α-Amylcinnamaldehyde dimethyl acetal

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

α-Pentylcinnamaldehyde dimethyl acetal, [2-(Dimethoxymethyl)-1-hepten-1-yl]benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Please contact Customer Service for Availability
CZK 4,340.00

About This Item

Linear Formula:
C6H5CH=C[(CH2)4CH3]CH(OCH3)2
CAS Number:
Molecular Weight:
248.36
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CZK 4,340.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

cinnamaldehyde

manufacturer/tradename

USP

refractive index

n20/D 1.508 (lit.)

bp

300 °C (lit.)

density

0.959 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC\C(=C/c1ccccc1)C(OC)OC

InChI

1S/C16H24O2/c1-4-5-7-12-15(16(17-2)18-3)13-14-10-8-6-9-11-14/h6,8-11,13,16H,4-5,7,12H2,1-3H3/b15-13+

InChI key

QCHZKUPVENJLAW-FYWRMAATSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

α-Amylcinnamaldehyde dimethyl acetal USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
103450110509491035005
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

refractive index

n20/D 1.508 (lit.)

refractive index

-

refractive index

n20/D 1.509 (lit.)

refractive index

n20/D 1.561 (lit.)

bp

300 °C (lit.)

bp

-

bp

83-85 °C/4 mmHg (lit.)

bp

234-237 °C (lit.)


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters