Skip to Content
Merck
All Photos(1)

Key Documents

EURM018

Human serum (antibodies against SARS-CoV-2)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 2,820.00

CZK 2,820.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CZK 2,820.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CZK 2,820.00


Please contact Customer Service for Availability

Request a Bulk Order

manufacturer/tradename

JRC

Quality Level

application(s)

clinical testing

storage temp.

−70°C

Analysis Note

For more information please click on the below product number:
EURM018

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service