Skip to Content
Merck
All Photos(1)

Key Documents

68197

Supelco

N-Nitrosopyrrolidine

analytical standard

Synonym(s):

1-Nitrosopyrrolidine, N-Nitrosopyrrolidine, NPYR

Sign Into View Organizational & Contract Pricing

Select a Size

68197-100MG
CZK 1,440.00

CZK 1,440.00


Please contact Customer Service for Availability


Select a Size

Change View
68197-100MG
CZK 1,440.00

About This Item

Empirical Formula (Hill Notation):
C4H8N2O
CAS Number:
Molecular Weight:
100.12
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

CZK 1,440.00


Please contact Customer Service for Availability

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.489 (lit.)

bp

214 °C (lit.)

density

1.085 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

O=NN1CCCC1

InChI

1S/C4H8N2O/c7-5-6-3-1-2-4-6/h1-4H2

InChI key

WNYADZVDBIBLJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Induces hepatocellular carcinomas and lung adenomas in mice. Forms DNA adducts that primarily result in A:T to G:C mutations.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service