Skip to Content
Merck
All Photos(1)

Key Documents

68088

Supelco

N-Nitrosomethylethylamine

analytical standard

Synonym(s):

N-Nitroso-N-methylethylamine, NMEA

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 3,730.00

CZK 3,730.00


Please contact Customer Service for Availability


Select a Size

Change View
100 MG
CZK 3,730.00

About This Item

Empirical Formula (Hill Notation):
C3H8N2O
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.24

CZK 3,730.00


Please contact Customer Service for Availability

grade

analytical standard

Quality Level

Assay

≥95%

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

N(N=O)(CC)C

InChI key

RTDCJKARQCRONF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.1 °F - closed cup

Flash Point(C)

76.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service