General description
Prevents bubbles and improves baseline stability. This 1/16 in. flange-type unit (4 × 1.3 cm) has a unique mechanism that ensures constant back-pressure over a wide range of mobile phase flows and viscosities. It is immune to clogging and other problems common to restrictor-type devices. Easily adjusts from 1-4 atm.(15-60 psi/1-4 kg/cm2).
Legal Information
SSI is a trademark of Scientific Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)