Skip to Content
Merck
All Photos(1)

Documents

22816

Porter Low Flow Pressure Regulator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500

General description

Provides precise pressure regulation at very low flows. Outlet pressure will not decrease more than 0.3 psi over the entire flow range.

Specifications
  • Regulation Range: 0-100 psig (0-7.0 kg/cm2).
  • Max. Operating Pressure: 250 psig (17.6 kg/cm2)
  • Pressure Drop Required: >10 psi (0.7 kg/cm2)
  • Control Accuracy: less than 0.3 psi decrease
  • Construction: aluminum body & bonnet, stainless steel diaphragm & filter, Buna-N O-rings, Viton valve seat

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wojciech Krzeptowski et al.
Antioxidants (Basel, Switzerland), 10(1) (2021-01-16)
G-quadruplexes (G4) are stacked nucleic acid structures that are stabilized by heme. In cells, they affect DNA replication and gene transcription. They are unwound by several helicases but the composition of the repair complex and its heme sensitivity are unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service