Skip to Content
Merck
All Photos(1)

Documents

575652-U

Supelco

SPE 96-Deep Square Well Collection Plate

well volume 1 mL, polypropylene, pk of 50

Synonym(s):

PK50 1ML PP 96 SQUARE WELL COLLECTION PLATES

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

material

polypropylene

packaging

pk of 50

technique(s)

solid phase extraction (SPE): suitable

well volume

1 mL

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This Sigma-Aldrich article discusses how the HybridSPE-Phospholipid Technology works and how the phospholipids are removed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service