Skip to Content
Merck
All Photos(1)

Key Documents

56700-U

Supelco

Ferrules

1/4-28 Upchurch for 1/16 in. O.D. tubing (requires Flangeless nut), ferrule only

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
CZK 370.00

CZK 370.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 EA
CZK 370.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

CZK 370.00


Please contact Customer Service for Availability

Request a Bulk Order

material

Tefzel®

packaging

pkg of 10 ea

manufacturer/tradename

Upchurch Scientific, Inc P-200X

fitting

1/4-28 Upchurch for 1/16 in. O.D. tubing (requires Flangeless nut)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Tefzel is a registered trademark of E. I. du Pont de Nemours and Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service