Key Documents
56629-U
Supelco
LC-MS Post Column Flow Splitters
Adjustable, Split Ratio = 1:1 to 20:1
Synonym(s):
LC-MS Flow Divider
Select a Size
1 EA
CZK 98,600.00
CZK 98,600.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
CZK 98,600.00
About This Item
UNSPSC Code:
41121815
NACRES:
SB.52
CZK 98,600.00
Please contact Customer Service for Availability
Request a Bulk Order
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)