Skip to Content
Merck

Skip To

48136

MISA Group 23 Chlorinated Hydrocarbon Mix

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
CZK 1,710.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

CZK 1,710.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

agency

MISA (Canada) Group 23

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

environmental
petroleum

format

multi-component solution

storage temp.

2-8°C

General description

MISA Group 23 Chlorinated Hydrocarbon Mix is an international and high-quality standard under MISA regulations, used as a reference standard for the routine monitoring of 11 chlorinated hydrocarbons, present in waste effluent samples.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4813348948CRM48140
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

multi-component solution

format

multi-component solution

format

multi-component solution

format

multi-component solution

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

300

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

grade

certified reference material, TraceCERT®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

concentration

2000 μg/mL each component in dichloromethane

concentration

2000 μg/mL each component in methanol

concentration

2000 μg/mL each component in methanol

concentration

2000 μg/mL in methanol


Analyte

Description

  • Hexachlorobenzene

  • Hexachloro-1,3-butadiene

  • Hexachlorocyclopentadiene

  • Hexachloroethane

  • Pentachlorobenzene

  • 1,2,3,4-Tetrachlorobenzene

  • 1,2,3,5-Tetrachlorobenzene

  • 1,2,4,5-Tetrachlorobenzene

  • 1,2,3-Trichlorobenzene

  • 1,2,4-Trichlorobenzene

  • 2,4,5-Trichlorotoluene

See All (11)

signalword

Danger

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-72-1 + 75-09-2 + 118-74-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters