Skip to Content
Merck
All Photos(2)

Documents

47205

Supelco

ASTM® D4815 Quantitative Calibration Mixture 1

with Internal Standard, (each component at the nominal wt% indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

ASTM® D4815

packaging

ampule of 1 mL

concentration

(each component at the nominal wt% indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

room temp

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Each component at the nominal weight percent listed.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    tert-Butanol .095 % (w/w)
    tert-Butyl methyl ether 19 % (w/w)
    1,2-Dimethoxyethane 5 % (w/w)
    Ethanol 2.85 % (w/w)
    2-Methyl-2-butanol 1.19 % (w/w)
    Isooctane:xylene (65:35) 71.87 % (w/w)

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-27.4 °F

Flash Point(C)

-33 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service