Skip to Content
Merck
All Photos(1)

Key Documents

47213

Supelco

ASTM® D4815 Qualitative Peak ID Mix

(each component at the nominal wt% indicated), analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
Pricing and availability is not currently available.

grade

analytical standard

Agency

ASTM® D4815

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

(each component at the nominal wt% indicated)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-30°C

General description

ASTM® D4815 Qualitative Peak ID Mix is for use in determination of oxygenates (Ethers and Alcohols) in Gasoline by GC.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Each component at the nominal weight percent listed. Use for determining relative retention times of selected oxygenates.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    tert-Amyl methyl ether 7.3 % (w/w)
    Benzene 5 % (w/w)
    1-Butanol 7.3 % (w/w)
    2-Butanol 7.3 % (w/w)
    tert-Butanol 7.3 % (w/w)
    tert-Butyl ethyl ether 4 % (w/w)
    tert-Butyl methyl ether 4 % (w/w)
    Diisopropyl ether 4 % (w/w)
    1,2-Dimethoxyethane 6 % (w/w)
    Ethanol 7.3 % (w/w)
    Methanol 7.3 % (w/w)
    2-Methyl-2-butanol 7.3 % (w/w)
    Methylcyclopentane 4 % (w/w)
    2-Methyl-1-propanol 7.3 % (w/w)
    1-Propanol 7.3 % (w/w)
    2-Propanol 7.3 % (w/w)
    See All (16)

Flash Point(F)

50.0 °F

Flash Point(C)

10 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service