Skip to Content
Merck
All Photos(1)

Key Documents

47129

Supelco

Cholestanol solution

certified reference material, 10 mg/mL in chloroform

Synonym(s):

(+)-Dihydrocholesterol solution, 3β-Cholestanol, 3β-Hydroxy-5α-cholestane, 5α-Cholestan-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H48O
CAS Number:
Molecular Weight:
388.67
Beilstein:
2418594
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in chloroform

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

140-142 °C (lit.)

application(s)

food and beverages

format

single component solution

storage temp.

2-30°C

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H]([C@H](C)CCCC(C)C)[C@@]4(C)CC[C@]3([H])[C@@]1(C)CC[C@H](O)C2

InChI

1S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28H,6-17H2,1-5H3/t19-,20+,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

QYIXCDOBOSTCEI-QCYZZNICSA-N

Gene Information

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michel R Hoenig et al.
Atherosclerosis, 184(2), 247-254 (2005-10-12)
Statins have been the mainstay of lipid-lowering therapy since their introduction. However, as lower LDL cholesterol targets are sought, adjunct therapies are becoming increasingly important. Few patients reach new targets with statin monotherapy. We propose that the cholestanol:cholesterol ratio can
Marja Lahti et al.
Environmental toxicology and chemistry, 31(8), 1738-1744 (2012-05-29)
The use of pharmaceuticals has increased enormously over the last few decades and serious concerns about their environmental fate and effects have arisen. Thus far, there is little knowledge about the historical pollution of the environment by pharmaceuticals. In the
Rozita Osman et al.
Environmental monitoring and assessment, 184(2), 1001-1014 (2011-04-16)
Increasing urbanization and changes in land use in Langat river basin lead to adverse impacts on the environment compartment. One of the major challenges is in identifying sources of organic contaminants. This study presented the application of selected chemometric techniques:
Claudia Gutterres Vilela et al.
Marine pollution bulletin, 62(10), 2055-2074 (2011-08-30)
Rodrigo de Freitas Lagoon, located in the Rio de Janeiro City, receives several types of polluted discharges. The knowledge of the sediment microfauna correlated with heavy metal and organic matter concentrations could supply important data about the conditions of the
Estelle Wolff et al.
The Journal of nutrition, 141(10), 1791-1798 (2011-08-26)
LDL-cholesterol (LDL-C) reduction may be achieved by various types of prudent diets, but their effects on surrogate markers of cholesterol absorption and synthesis have not been well studied in humans. We aimed to assess whether the extent of cholesterol absorption

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service