Skip to Content
Merck

Skip To

W0765

Well plate stand

pkg of 1 Stand

Synonym(s):

multiplate stand, Diversified Biotech WPST-1000, plate holder, plate rack, plate support

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
CZK 1,450.00

About This Item

NACRES:
NB.22
UNSPSC Code:
41122102
Manufacturer/tradename:
Diversified Biotech WPST-1000
Packaging:
pkg of 1 Stand

CZK 1,450.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

(tips the Well Orienter card and the microplate to a 30° angle)

sterility

non-sterile

packaging

pkg of 1 Stand

manufacturer/tradename

Diversified Biotech WPST-1000

color

none

General description

This device tips the Well Orienter card and the microplate 30° allowing the investigator to identify wells clearly whether sitting or standing. Comfortably view and pipet without strain. The Well Stand will free up one hand since you do not have to constantly tip the plate back and forth while pipetting.
Each stand has a neoprene pad and rubber mounting to prevent slippage of the plate and stand.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z743557Z363359HS120480
manufacturer/tradename

Diversified Biotech WPST-1000

manufacturer/tradename

Diversified Biotech WPAJ-1000

manufacturer/tradename

Bel-Art F18938-0000

manufacturer/tradename

Heathrow Scientific HS120480

packaging

pkg of 1 Stand

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

-

sterility

non-sterile

sterility

-

sterility

non-sterile

sterility

-

description

(tips the Well Orienter card and the microplate to a 30° angle)

description

(tips the Well Orienter card and the microplate to a 30° angle)

description

-

description

-

color

none

color

-

color

none

color

blue to green, 8 of each color for top + 8 blue bottoms


Still not finding the right product?

Explore all of our products under Well plate stand




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
W0765-1EA04061826747810