Skip to Content
Merck

Skip To

U7883

Monoclonal Anti-USP25 antibody produced in mouse

~1.0 mg/mL, clone U25.7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-USP21, Anti-Ubiquitin specific peptidase 25

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
U25.7, monoclonal
Application:
ELISA (i), ICC, WB
Citations:
2
To inquire about this product U7883, please contact your local Merck office or dealer. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

U25.7, monoclonal

form

buffered aqueous solution

mol wt

antigen ~126 kDa

species reactivity

rat, mouse, human

concentration

~1.0 mg/mL

technique(s)

immunocytochemistry: suitable, indirect ELISA: suitable, western blot: 1-2 μg/mL using total extract of HEK-293T cells expressing USP25.

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP25(29761)
mouse ... Usp25(30940)
rat ... Usp25(304150)

General description

Monoclonal Anti-USP25 (mouse IgG2a isotype) is derived from the hybridoma U25.7 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to a fragment of human USP25. USP25 (ubiquitin-specific protease 25) is a member of the ubiquitin processing proteases (UBP) family. It is located on the gene-poor region of 21q11.2 human chromosome.

Application

Monoclonal Anti-USP25 antibody has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting
  • immunocytochemistry

Biochem/physiol Actions

USP25 (ubiquitin-specific protease 25) is a target for sumoylation. It takes place within the two ubiquitin interaction motifs (UIM) of USP25 that are required for efficient hydrolysis of Ub (ubiquitin), and thus impairs USP25 activity as a result of reduced affinity to Ub chains. Overexpression of USP25 has been observed in Down syndrome fetal brain.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
WH0027005M3WH0057663M1HPA018297
clone

U25.7, monoclonal

clone

3D10, monoclonal

clone

1A8, monoclonal

clone

polyclonal

Gene Information

human ... USP25(29761)
mouse ... Usp25(30940)
rat ... Usp25(304150)

Gene Information

human ... USP21(27005)

Gene Information

human ... USP29(57663)

Gene Information

human ... USP25(29761)

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit

species reactivity

rat, mouse, human

species reactivity

human

species reactivity

human

species reactivity

human


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions


Erik Meulmeester et al.
Molecular cell, 30(5), 610-619 (2008-06-10)
Vertebrates express two distinct families of SUMO proteins (SUMO1 and SUMO2/3) that serve distinct functions as posttranslational modifiers. Many proteins are modified specifically with SUMO1 or SUMO2/3, but the mechanisms for paralog selectivity are poorly understood. In a screen for
A Bosch-Comas et al.
Cellular and molecular life sciences : CMLS, 63(6), 723-734 (2006-02-28)
The biological functions of the more than one hundred genes coding for deubiquitinating enzymes in the human genome remain mostly unknown. The USP25 gene, located at 21q11.2, encodes three protein isoforms produced by alternative splicing. While two of the isoforms



Global Trade Item Number

SKUGTIN
U7883-200UL04061837898310

Questions

Reviews

No rating value

Active Filters