Skip to Content
Merck

Skip To

U5010

UNISETTE Biopsy Processing/Embedding Cassettes

yellow

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NB.17
UNSPSC Code:
12352202


color

yellow

application(s)

hematology
histology

storage temp.

room temp

General description

Histology processing/embedding cassettes made of acetal polymer. These cassettes keep specimens safely submerged in liquid and are resistant to histological solvents. The one-piece lid snaps apart and conveniently locks into the cassette base, ensuring specimen safety. A line of cassette colors are available to fit your needs and preference. A large 30° anterior writing area is available for your convenience. Not suitable for automated printers.

Application

Compatible with Leica and Sakura printers.

Preparation Note

Case contains 3 dispenser boxes of 500 cassettes.

Legal Information

Unisette is a trademark of Simport


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
U5010-1CS04061826763551

Questions

Reviews

No rating value

Active Filters