Key Documents
TMO312089-0004
Nalgene® narrow-mouth bottle
natural high-density polyethylene, size 125 mL, case of 500 ea
Synonym(s):
Nalgene bottle, plastic bottle
Select a Size
500 EACH
CZK 12,800.00
CZK 12,800.00
Please contact Customer Service for Availability
Select a Size
Change View
500 EACH
CZK 12,800.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
CZK 12,800.00
Please contact Customer Service for Availability
material
natural high-density polyethylene
packaging
case of 500 ea
manufacturer/tradename
Nalgene 312089-0004
size
125 mL
General description
Store and ship light-sensitive reagents with Thermo Scientific Nalgene Narrow-Mouth Opaque Amber HDPE Packaging Bottles with Closure. Ideal for diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals, these high density polyethylene bottles are injection blow-molded for uniform wall thickness and strength. They meet USP light transmission standards and are bulk packed for packaging applications.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)