Skip to Content
Merck

Skip To

T6580

Triflusal

≥98% (HPLC), powder

Synonym(s):

Triflusal, α,α,α-trifluoro-2,4-cresotic acid acetate, 2-(Acetyloxy)-4-(trifluoromethyl)benzoic acid, Drisgen, acetyl-4-trifluoromethylsalicylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H7F3O4
CAS Number:
Molecular Weight:
248.16
UNSPSC Code:
12352200
EC Number:
206-297-5
NACRES:
NA.25
Assay:
≥98% (HPLC)
Form:
powder
To inquire about this product T6580, please contact your local Merck office or dealer. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >30 mg/mL

originator

Uriach

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(c(cc1)C(=O)O)OC(=O)C

InChI

1S/C10H7F3O4/c1-5(14)17-8-4-6(10(11,12)13)2-3-7(8)9(15)16/h2-4H,1H3,(H,15,16)

InChI key

RMWVZGDJPAKBDE-UHFFFAOYSA-N

Biochem/physiol Actions

Triflusal is an anti-inflammatory antithrombotic platelet aggregation inhibitor with a 3-fold mechanism of action.
Triflusal is an anti-inflammatory antithrombotic platelet aggregation inhibitor with a 3-fold mechanism of action. It is an irreversible inhibitor of COX-1 inhibiting thromboxane A2, preventing aggregation; it preserves vascular prostacyclin, thus promoting anti-aggregant effect. Triflusal blocks phosphodiesterase leading to an increase in cAMP levels promoting anti-aggregant effect due to inhibition of calcium mobilization.

Features and Benefits

This compound was developed by Uriach. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2841H0131N0630
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

solubility

DMSO: >30 mg/mL

solubility

DMSO: 2 mg/mL, clear

solubility

H2O: ~50 mg/mL

solubility

acetone: 50 mg/mL, clear to slightly hazy, yellow

originator

Uriach

originator

-

originator

-

originator

-


Still not finding the right product?

Explore all of our products under Triflusal


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.






Global Trade Item Number

SKUGTIN
T6580-5MG04061833027417
T6580-25MG04061833027400

Questions

Reviews

No rating value

Active Filters