Merck
Skip To
T5540
Tweezers
Style #3, very sharp, PTFE-coated, anti-magnetic stainless steel
Synonym(s):
fine forceps, fine tweezers, needle tweezers, Ted Pella 5320, laboratory forceps
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
CZK 1,670.00
About This Item
NACRES:
MA.01
UNSPSC Code:
27112105
CZK 1,670.00
Please contact Customer Service for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
PTFE-coated, anti-magnetic stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Ted Pella 5320
size
120 mm
General description
Specifically designed for microscopy applications. All are ultra-high precision tools of Swiss manufacture.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
T6040T5790T4287
Tweezers Style #3, very sharp, PTFE-coated, anti-magnetic stainless steel
T5540
Tweezers
Tweezers Style #5, Fine needle-sharp, PTFE-coated, anti-magnetic stainless steel
T6040
Tweezers
Tweezers Style #4, needle-sharp, anti-magnetic stainless steel
T5790
Tweezers
Tweezers Style #3C, very sharp, anti-magnetic stainless steel
T4287
Tweezers
material
PTFE-coated, anti-magnetic stainless steel
material
PTFE-coated, anti-magnetic stainless steel
material
anti-magnetic stainless steel
material
anti-magnetic stainless steel
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
manufacturer/tradename
Ted Pella 5320
manufacturer/tradename
Ted Pella 5622-TEF
manufacturer/tradename
Ted Pella 504-NM
manufacturer/tradename
Ted Pella 503C-NM
size
120 mm
size
110 mm
size
110 mm
size
110 mm
Still not finding the right product?
Explore all of our products under Tweezers
Choose from one of the most recent versions:
Certificates of Analysis (COA)