Merck
Skip To
T0943
Pharmed L/S tubing
size 17
Synonym(s):
Saint Gobain AY242019, PeristalticPump Tubing
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Pack SizeSKUAvailabilityPrice
25 ft
Please contact Customer Service for Availability
CZK 8,100.00
About This Item
NACRES:
NB.35
UNSPSC Code:
41105108
CZK 8,100.00
Please contact Customer Service for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pack of 25 feet
manufacturer/tradename
Tygon
I.D.
0.25 in. (6.4 mm)
General description
Tubing for MasterFlex Peristaltic Pumps, PharMed tubing has been specifically formulated for the MasterFlex pump system. Great for tissue and cell work- nontoxic and nonhemolytic. Long service life minimizes risk of fluid exposure; reduces tubing cost and pump downtime.
  • Opaque to UV and visible light to protect light-sensitive fluids
  • Heat sealable, bondable, and formable
  • Extremely low gas permeability
Legal Information
MasterFlex is a trademark of Cole-Parmer Instrument Co.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
T0568T0443T1068
I.D.
0.25 in. (6.4 mm)
I.D.
0.06 in. (1.6 mm)
I.D.
0.03 in. (0.8 mm)
I.D.
0.31 in. (7.9 mm)
packaging
pack of 25 feet
packaging
pack of 25 feet
packaging
pack of 25 feet
packaging
pack of 25 feet
manufacturer/tradename
Tygon
manufacturer/tradename
Tygon
manufacturer/tradename
Tygon
manufacturer/tradename
Tygon
Still not finding the right product?
Explore all of our products under Pharmed L/S tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Global Trade Item Number
SKUGTIN
T0943-25FT04061834381549