Skip to Content
Merck

Skip To

T0924

2,3,4-Tri-O-benzyl-L-fucopyranose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H30O5
CAS Number:
Molecular Weight:
434.52
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

−20°C

SMILES string

C[C@@H]1OC(O)[C@@H](OCc2ccccc2)[C@H](OCc3ccccc3)[C@@H]1OCc4ccccc4

InChI

1S/C27H30O5/c1-20-24(29-17-21-11-5-2-6-12-21)25(30-18-22-13-7-3-8-14-22)26(27(28)32-20)31-19-23-15-9-4-10-16-23/h2-16,20,24-28H,17-19H2,1H3/t20-,24+,25+,26-,27?/m0/s1

InChI key

YRAQXZMHYZXWBZ-OGPNLZSQSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library